Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD324 (E-Cadherin) Mouse anti-Human, BV480, Clone: 67A4  , BD Horizon™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 565646

Catalog No. BDB565646

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

CD324 (E-Cadherin) Monoclonal antibody specifically detects CD324 (E-Cadherin) in Human samples. It is validated for Immunofluorescence.
Specifications

Specifications

CD324 (E-Cadherin)
Monoclonal
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
50 μg
Cell Biology
Human
Purified
Immunofluorescence
67A4  
Brilliant Violet 480
E-cadherin; CD324; CDH1; CADH1; Cadherin-1; ECAD; CDHE; Arc-1; LCAM; UVO
Human Breast Tumor Cell Line
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed.
Mouse BALB/c IgG1 κ
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.