Learn More
CD33 Mouse anti-Human, Clone: OTI7G8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806758
Catalog No. 50-166-7960
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.
Specifications
Specifications
CD33
Monoclonal
Unconjugated
CD33
Lyophilized
CD33
Full length human recombit protein of human CD33 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI7G8
PBS with 8% trehalose and no preservative; pH 7.3
P20138
SIGLEC-3, SIGLEC3, p67
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
945