Learn More
CD33 Mouse anti-Human, FITC, Clone: WM53, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15763
Catalog No. 50-113-7753
Add to cart
Description
Description
CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.
Specifications
Specifications
CD33, CD33
Monoclonal
FITC
CD33
Liquid
CD33
Human AML cells
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
WM53
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
P20138
FLJ00391, SIGLEC-3, p67
Mouse
IgG1
100 tests
4° C
Primary
945