Learn More
CD33 Mouse, anti-Human, R718, Clone: P67.6, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751936
Catalog No. BDB751936
Add to cart
Description
Description
The monoclonal antibody specifically recognizes CD33, a human myelomonocytic antigen which is also known as Sialic acid-binding Ig-like lectin 3 (Siglec-3 or SIGLEC3). CD33 is a 67 kDa type I transmembrane glycoprotein that belongs to the Ig supergene family. The CD33 antigen is present on monocytes (bright) and granulocytes (dim). Granulocytes can be further subdivided into neutrophil, eosinophil, and basophil populations based on CD33 staining in combination with other cell-surface antigens. The CD33 antigen is also found on CFU-Mix, CFU-GM, CFU-Meg, a portion of BFU-E, myeloblasts, promyelocytes, myelocytes, and metamyelocytes, but not on earlier precursors. The CD33 antigen is expressed on blast cells in greater than 85% of acute myeloid leukemias (AML), and it can be aberrantly expressed in acute lymphoblastic leukemias (ALL). Normal lymphocytes, platelets, and erythrocytes do not express the CD33 antigen. CD33 can reportedly function as a sialic acid-dependent cell adhesion molecule and this function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane.
Specifications
Specifications
CD33
P67.6
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human