Learn More
CD335 (NKp46) Monoclonal Antibody (9E2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$228.00 - $435.00
Specifications
| Antigen | CD335 (NKp46) |
|---|---|
| Clone | 9E2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Description: This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type I diabetes. The 9E2 antibody has been reported to induce cytotoxicity. Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human periheral blood cells. This can be used at 5 μL (0.03 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat.
NKp46 (CD335, NCR1, ) is a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. NKp46 consists of two Ig-like domains assembled to leukocyte immunoglobulin-like (LIR) and killer inhibitory receptors (KIR).Specifications
| CD335 (NKp46) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
| NCR1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| NCR1 |
| 9E2 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Mouse | |
| Human | |
| O76036 | |
| 9437 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.