Learn More
CD338 (ABCG2) Mouse anti-Human, PE, Clone: 5D3, eBioscience™ Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Catalog No. 12-888-841
Add to cart
Description
Description
The 5D3 MAb reacts with the extracellular portion of the human ABCG2 protein, also known as Bcrp1 and MXR. The ABCG2 gene, a member of the multi-drug resistance (MDR) family, is highly expressed on primitive 'side-population' (SP) stem cells, which are defined by the efflux of fluorescent dyes such as Rhodamine 123 and Hoechest 33342. In the bone marrow, about 0.05% of cells display the low fluorescence and are highly enriched for repopulating cells. The SP cells, which express low or undetectable levels of CD34, have been identified in multiple species. In addition, expression of ABCG2 appears to be highly conserved.
CD338 (ABCG2) Monoclonal antibody specifically detects CD338 (ABCG2) in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
Liquid
Flow Cytometry
5D3
PE
ABCG2
ABC transporter; ABC15; ABCG2; ABCP; ATP binding cassette subfamily G member 2 (Junior blood group); ATP-binding cassette sub-family G member 2; ATP-binding cassette transporter G2; ATP-binding cassette, sub-family G (WHITE), member 2 (Junior blood group); BCRP; BCRP1; BMDP; Breast cancer resistance protein; Broad substrate specificity ATP-binding cassette transporter ABCG2; CD338; CDw338; EST157481; GOUT1; Mitoxantrone resistance-associated protein; MRX; multi drug resistance efflux transport ATP-binding cassette sub-family G (WHITE) member 2; MXR; MXR1; MXR-1; placenta specific MDR protein; placenta-specific ATP-binding cassette transporter; UAQTL1; Urate exporter
IgG2b, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
9429
CD338 (ABCG2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9UNQ0
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human