Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Antibody (OTI11E1), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270375FR

Catalog No. NBP270375FR


Add to Cart

Description

Description

CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CD34
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
CD34
Recombinant protein fragment corresponding to amino acids 312-385 of human CD34 (NP_001020280) produced in E.coli.
0.1 mL
Primary
Human
IgG2a
Western Blot, Immunohistochemistry
OTI11E1
Western Blot, Immunohistochemistry
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Mouse
Affinity Purified
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
947.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit