Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CD34 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
$1219.00
Specifications
Assay Range | 156.25 pg/mL |
---|---|
Assay Sensitivity | 156.25-10000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB010229
|
Novus Biologicals™
NBP27959515PLATES |
15 Plates |
Each of 1 for $1,219.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse CD34.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse CD34. | |
Sandwich ELISA | |
947 | |
Rabbit | |
RUO | |
Storage is content dependent. |
156.25-10000 pg/mL | |
HRP | |
Antibody Pairs | |
Mouse | |
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
CD34 | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
CD34 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title