Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI11E1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270375AF405

Catalog No. N270375A405


Add to cart

Description

Description

CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CD34
OTI11E1
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
CD34
Recombinant protein fragment corresponding to amino acids 312-385 of human CD34 (NP_001020280) produced in E.coli.
Immunogen affinity purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
Primary
947.0
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit