Learn More
CD34 Mouse anti-Human, Clone: 581 (PE), Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13879
Catalog No. 89-033-944
Add to cart
Description
Description
Mouse monoclonal antibody raised against human CD34.
  • High specificity
  • High reactivity
Specifications
Specifications
CD34
581
Mouse monoclonal antibody raised against human CD34.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
Mouse
IgG1, kappa
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Flow Cytometry
PE
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
P28906
CD34
Human CD34+ leukemic cells.
Protein A/G purification
RUO
Primary
947
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)