Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant human CD34.

Manufacturer:  Abnova Corporation PAB29566

Catalog No. 89-933-791


Add to cart

Description

Description

CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110 kD that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM]

Sequence: ATSPTKPYTSSSPILSDIKAEIKCSGIREVKLTQGICLEQNKTSSCAEFKKDRGEGLARVLCGEEQADADAG
Specifications

Specifications

CD34
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Liquid
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against recombinant human CD34.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
CD34
Recombinant protein corresponding to amino acids of human CD34.
Antigen affinity purification
RUO
Primary
947
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit