Learn More
CD34 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS2038R
Encompass_Preferred
Catalog No. 50-198-4453
Add to cart
Description
Description
CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
Specifications
Specifications
CD34
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Cd34
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
947
Cd34
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P28906
AU040960; CD34; CD34 antigen; CD34 molecule; cluster designation 34; Hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; LOC100343670; Mucosialin; RP11-328D5.2
KLH conjugated synthetic peptide derived from mouse CD34.
Protein A
RUO
Antibody
Polyclonal
Human