Learn More
CD34 Rabbit anti-Human,Rat, KHL, Clone: 5C15, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB123225UL
 View more versions of this product
Catalog No. ZRB123225MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD34 Monoclonal antibody specifically detects CD34 in Human,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
CD34
5C15
KHL
Purified
CD34
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Hematopoietic progenitor cell antigen CD34.
Target Molecular Weight 40.72 kDa calculated; ∼110 kDa observed. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_001020280.1
Hematopoietic progenitor cell antigen CD34
Rabbit
IgG
Protein A Purified
Inflammation and Immunology
Primary
Human, Rat