Learn More
CD340 (HER-2) Mouse anti-Human, BV421, Clone: 24D2, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566459
 View more versions of this product
Catalog No. BDB566459
Add to cart
Description
Description
CD340 (HER-2) Monoclonal antibody specifically detects CD340 (HER-2) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD340 (HER-2)
24D2
Brilliant Violet 421
Mouse
25 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human