Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Antibody (594708), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5748H
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CD35 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 594708 | |
| Flow Cytometry, CyTOF-ready | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Mouse myeloma cell line NS0-derived recombinant human CD35, Gln42-Asp1971 (His1208Arg), Accession # CAA68755 | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction