Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB515, Clone: E11, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566021
Description
CD35 Monoclonal antibody specifically detects CD35 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.Specifications
CD35 | |
Monoclonal | |
BB515 | |
CR1; Complement receptor type 1; C3b/C4b receptor; C3BR; C4BR; KN | |
Human Cells of the Monocyte Lineage | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
Mouse IgG1 κ |
Flow Cytometry | |
E11 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.