Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Rabbit anti-Human, Clone: CR1/4382R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen CD35
Clone CR1/4382R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122979
View Documents
Novus Biologicals
NBP30775310UG
100 μg
Each for $406.52
Add to cart
 
NB122980
View Documents
Novus Biologicals
NBP30775320UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

CD35
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
1378
IgG
Primary
Store at 4°C.
CR1/4382R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
complement component (3b/4b) receptor 1 (Knops blood group), KN
Synthetic peptide corresponding to CD35 residues within aa1939-2039 of CD35 was used as an immunogen.
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit