Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD35 Rabbit anti-Mouse, Alexa Fluor™ 750, Clone: 7G6, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252667AF750

Catalog No. NP22667A70


Add to cart

Description

Description

CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications

Specifications

CD35
7G6
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
complement component (3b/4b) receptor 1 (Knops blood group), KN
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Recognises an epitope shared by 145-150kDa and 190kDa complement receptor proteins, originally designated CR2 (CD21) and CR1 (CD35).
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CR1
Purified Mouse CR1.
0.1 mL
Primary
1378
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit