Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD35 Rabbit anti-Mouse, Clone: 7G6, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 1 publication
$229.00 - $516.00
Specifications
Antigen | CD35 |
---|---|
Clone | 7G6 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25266725
|
Novus Biologicals
NBP2526670.025MG |
0.025 mg |
Each for $229.00
|
|
NBP25266702
|
Novus Biologicals
NBP2526670.2MG |
0.2 mg |
Each for $516.00
|
|
Description
CD35 Monoclonal specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready, Block/Neutralize.Specifications
CD35 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
1378 | |
Purified Mouse CR1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
7G6 | |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Mouse | |
complement component (3b/4b) receptor 1 (Knops blood group), KN | |
CR1 | |
IgG | |
Protein A or G purified | |
Recognises an epitope shared by 145-150kDa and 190kDa complement receptor proteins, originally designated CR2 (CD21) and CR1 (CD35). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title