Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD36 Mouse anti-Human, PE, Clone: NL07, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen A15793

Catalog No. 50-113-7767


Add to cart

Description

Description

CD36 (fatty acid translocase, FAT) is an 88 kDa, ditopic glycosylated protein that belongs to the class B family of scavenger receptors. CD36 is expressed by most resting marginal zone B cells but not by follicular and B1 B cells, and is rapidly induced on Follicular B cells in vitro upon TLR and CD40 stimulation. CD36 does not affect the development of B cells, but modulates both primary and secondary antibody response. Similar to glucose transporter GLUT4, CD36 is translocated from intracellular pools to the plasma membrane following cell stimulation by insulin. In mouse, CD36 is responsible for gustatory perception of long-chain fatty acids. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signaling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. Further, CD36 may function as a cell adhesion molecule and directly mediates cyto-adherence of Plasmodium falciparum parasitized erythrocytes. Mutations in the CD36 gene cause platelet glycoprotein deficiency. Multiple alternatively spliced transcript variants encoding the CD36 protein have been found.
Specifications

Specifications

CD36
Monoclonal
PE
CD36
Liquid
CD36
Human CD36
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
NL07
PBS with 0.2% BSA and 15mM sodium azide
P16671
FAT, GPIV, Scarb3, GPIIIB, PAS IV, PAS-4, fatty acid translocase, glycoprotein IIIb, platelet glycoprotein 4, platelet glycoprotein IV
Mouse
IgM
100 tests
4° C
Primary
948
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit