Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD367 (Clec4A, DCIR) Mouse anti-Human, PE, Clone: 9E8, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 12987542

Catalog No. 50-112-2484


Add to cart

Description

Description

Description: This 9E8 monoclonal antibody reacts with human CD367, also known as dendritic cell immunoreceptor (DCIR) or Clec4A. This C-type lectin is expressed on monocytes, granulocytes, B cells, and plasmacytoid dendritic cells in peripheral blood. DCIR is also expressed on mature and immature monocyte-derived dendritic cells in vitro. DCIR is not expressed on NK or T cells. With its cytoplasmic ITIM domain, DCIR is an inhibitory receptor that recruits the SHP-1 and SHP-2 phosphatases upon ligand binding. Studies have demonstrated that DCIR plays a role in the interaction between dendritic cells and HIV-1. Applications Reported: This 9E8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E8 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in inflammatory and immune response. Multiple transcript variants encoding distinct isoforms have been identified for this gene. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications

Specifications

CD367 (Clec4A, DCIR)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9UMR7
CLECSF6, DCIR, DDB27, LLIR, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6, C-type lectin DDB27, C-type lectin domain family 4 member A, C-type lectin superfamily member 6, dendritic cell immunoreceptor, lectin-like immunoreceptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
9E8
PE
CLEC4A
Liquid
CLEC4A
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
50856
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.