Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD37 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS541PABX

 View more versions of this product

Catalog No. MS541PABX

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

CD37, a tetraspan transmembrane protein, is strongly expressed on normal and neoplastic mature (sIg+) B lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD37 is absent from platelets and erythrocytes. It associates with other tetraspan transmembrane proteins and MHC class II molecules to form a large complexes at the surface of B cells and play a role in signal transduction. CD37 is a valuable and stable marker for peripheral mature B-cells and corresponding malignancies like B-cell chronic lymphocytic leukemia (B-CLL), hairy cell leukemia (HCL), and all types of B-cell non-Hodgkin lymphoma (B-NHL).

Host Species: Mouse

Clone: IPO-24

Isotype: IgG2b/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Spleen cells of a patient with hairy cell leukemia

Molecular Weight: 33-55 kDa

Positive Control: Raji cells, Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD37 Ab-1
Monoclonal
1mg/mL
Concentrated
Spleen cells of a patient with hairy cell leukemia
200μL
B-Cell
Primary
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
IPO-24
Unconjugated
Murine
IgG2b kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit