Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD371 (Clec12A) Mouse anti-Human, BV480, Clone: 50C1, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 746824
Description
CD371 (Clec12A) Monoclonal antibody specifically detects CD371 (Clec12A) in Human samples. It is validated for Flow Cytometry.Specifications
CD371 (Clec12A) | |
50C1 | |
Brilliant Violet 480 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG2a, κ | |
RUO | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title