Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Antibody (OTI1C9), HRP, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270376H

Catalog No. NBP270376H


Add to Cart

Description

Description

CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

CD38
Monoclonal
HRP
PBS with No Preservative
CD38
Recombinant protein fragment corresponding to amino acids 43-300 of human CD38 (NP_001766) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI1C9
Western Blot, Immunohistochemistry
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Mouse
Affinity Purified
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers
952.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit