Learn More
AdipoGenSupplier Diversity Partner CD38 (HU):FC(MS)(REC.)(100UG) DFS Item
Manufacturer:  AdipoGenSupplier Diversity Partner CHIHF211CD38C100
CD38 (human):Fc (mouse) (rec.), AdipoGen Life Sciences. Protein. The ed of human CD38 (aa 43 - 300) is fused to the N-terminus of the Fc region of mouse IgG2a. Lyophilized from 0.2�m-filtered solution in PBS.
Catalog No. 50-114-7391