Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Antibody (HB7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP252696AF350
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD38 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CD38 | |
Viable cells (1 x 10E7) of the human cell line, BJAB (Human Burkitt lymphoma B cell line), which was were injected intraperitoneally into 8-week-old BALB/c mice. | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
952 | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry | |
HB7 | |
Western Blot, Flow Cytometry | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
Mouse | |
0.1 mL | |
Primary | |
This antibody binds to an epitope between amino acids 273-285 of human CD38, an approximately 45 kDa type II transmembrane protein, expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. Also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. Bimodally expressed during B cell development, modulating from high in immature cells to low in intermediate ones and back to high on mature B cells. This antibody competes with clone AT13/5 (Ab00289) (Ellis 1995). | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction