Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Mouse anti-Human, APC, Clone: HB7 (also known as HB-7), BD Pharmingen™
Mouse Monoclonal Antibody
$130.00 - $285.00
Specifications
Antigen | CD38 |
---|---|
Clone | HB7 (also known as HB-7) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow CytometrySpecifications
CD38 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; OKT10 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
HB7 (also known as HB-7) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human BJAB B cell line | |
Primary |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title