Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Mouse anti-Human, BB700, Clone: HIT2, BD Horizon™

Mouse Monoclonal Antibody

$190.00 - $385.00

Specifications

Antigen CD38
Applications Flow Cytometry
Host Species Mouse
Target Species Human
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566446
View Documents
BD Biosciences
566446
25 Tests Each for $190.00
Add to cart
 
BDB566445
View Documents
BD Biosciences
566445
100 Tests Each for $385.00
Add to cart
 
Description

Description

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD38
Mouse
Primary
Flow Cytometry
Human
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit