Learn More
CD38 Mouse anti-Human, Clone: GR7A4 (APC-C750), Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13893
Catalog No. 89-033-958
Add to cart
Description
Description
Mouse monoclonal antibody raised against human CD38.
  • High specificity
  • High reactivity
Specifications
Specifications
CD38
GR7A4(LD38)
Mouse monoclonal antibody raised against human CD38.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
CD38
Human acute lymphoblastic leukemia (ALL) B cells.
Protein A/G purification
RUO
Primary
952
Flow Cytometry
Unconjugated
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
P28907
T10
Mouse
IgG1, kappa
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)