Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1C9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270376JF646

Catalog No. N270376J646


Add to cart

Description

Description

CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CD38
OTI1C9
Immunohistochemistry
Affinity Purified, Conjugated
CD38
Recombinant protein fragment corresponding to amino acids 43-300 of human CD38 (NP_001766) produced in E.coli.
Immunogen affinity purified
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers
Primary
952.0
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit