Learn More
CD38 Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: 1G7F4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225250AF532
Catalog No. NP2220A32
Add to cart
Description
Description
CD38 Monoclonal antibody specifically detects CD38 in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
CD38
1G7F4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10
Mouse
IgG2b Lambda
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD38
Amino acids 71-197 of mouse CD38 were used as the immunogen for the antibody.
0.1 mL
Primary
952