Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Rabbit anti-Human, Clone: 6F10, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA538376

Encompass

Catalog No. PIMA538376


Add to cart

Description

Description

This antibody has been tested in direct-ELISA

CD38 Recombinant Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

CD38
Recombinant Monoclonal
0.27 mg/mL
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide
P28907
2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10
A synthesized peptide derived from human CD38
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
6F10
Unconjugated
CD38
Liquid, Affinity Purified
Rabbit
IgG
100 μL
-20°C or -80°C if preferred
Primary
952
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit