Learn More
CD39/ENTPD1 Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 350, Clone: 17B5F11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225223AF350
Catalog No. NP22223A30
Add to cart
Description
Description
CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CD39/ENTPD1
17B5F11
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
Mouse
IgG2b Lambda
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ENTPD1
Amino acids 102-208 of human CD39 were used as the immunogen for the antibody.
0.1 mL
Primary
953