Learn More
CD39 Mouse anti-Human, Alexa Fluor® 647, Clone: A1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD3921
Catalog No. 50-112-1406
Add to cart
Description
Description
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Specifications
Specifications
CD39
Monoclonal
Alexa Fluor™ 647
ENTPD1
Liquid
ENTPD1
Human CD39
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
A1
PBS with BSA and 0.1% sodium azide
P49961
ATPDase, CD39, DKFZp686D194, DKFZp686I093, FLJ40921, FLJ40959, NTPDase-1
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
953