Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD39 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547624
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CD39 and less than 1% cross-reactivity with rhCD39L2, rhCD39L3 and recombinant mouse CD39L3. Reconstitute at 0.2 mg/mL in sterile PBS.
CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Specifications
CD39 | |
Polyclonal | |
Unconjugated | |
ENTPD1 | |
2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
12495 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P55772 | |
ENTPD1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD39/ENTPD1 Thr38-Ile478. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction