Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD39L3/ENTPD3 Human Recombinant Protein, TMW: 52kDa

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Manufacturer:  Novus Biologicals™ NBP2519470.01MG

 View more versions of this product

Catalog No. NBP25947001


Add to cart

Description

Description

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 44-485 of Human CD39L3/ENTPD3 The Recombinant Human CD39L3/ENTPD3 Protein is derived from E. coli. The Recombinant Human CD39L3/ENTPD3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
TMW: 52kDa
0.01mg
CD39 antigen-like 3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, FLJ93839, HB6NTPDase 3, NTPDase-3
Unlabeled
SDS-Page
956
>85%, by SDS-PAGE
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CD39L3/ENTPD3
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis