Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD39L4/ENTPD5 Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793505PLATES

 View more versions of this product

Catalog No. NB009740


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD39L4/ENTPD5.
Specifications

Specifications

23.4 pg/mL
Sandwich ELISA
  • Standard: Each vial contains 27 ng of recombinant Human CD39L4/ENTPD5.
  • Capture Antibody: 0.5 mg/mL of rabbit anti-Human CD39L4/ENTPD5 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human CD39L4/ENTPD5 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • RUO
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD39L4/ENTPD5.
    CD39L4/ENTPD5
    Lipid and Metabolism
    Human
    ENTPD5
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
    HRP
    957
    Antibody Pairs
    23.4-1500 pg/mL
    Storage is content dependent.
    Rabbit
    5 Plates
    Sandwich ELISA
    CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5
    SDS
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit