Learn More
Abnova™ CD3D 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000915T02
Catalog No. 89-938-695
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CD3D 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 18.9kDa
Transient overexpression cell lysate was tested with anti-CD3D Purified antibody (Mfr. No. H00000915-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_000732
pCMV-CD3D full-length
100 μL
Human