Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd3e, anti-Mouse, Clone: 145-2C11, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00108

Catalog No. 89-032-451


Add to cart

Specifications

Specifications

Cd3e
145-2C11
Flow Cytometry (1 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
CD3 antigen, epsilon polypeptide
AI504783|CD3|CD3epsilon|T3e
Original antibody is raised against H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
12501
Mouse
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
Cd3e
IgG, kappa
RUO
Antibody
Monoclonal
This antibody recognizes a 25 kDa protein component of the antigen-specific T-cell receptor.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit