Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3E Hamster anti-Mouse, PerCP-Cy5.5, Clone: 145-2C11, Abnova™
Armenian Hamster monoclonal antibody raised against Cd3e.
Supplier: Abnova Corporation MAB12205
Specifications
CD3E | |
Monoclonal | |
PerCP-Cyanine5.5 | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
AI504783/CD3/CD3epsilon/T3e | |
Hamster | |
Affinity Chromatography | |
RUO | |
12501 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
IgG |
Flow Cytometry | |
145-2C11 | |
Armenian Hamster monoclonal antibody raised against Cd3e. | |
In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
Cd3e | |
Cd3e | |
100 μg | |
Primary | |
Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction