Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3E Mouse anti-Human, Clone: 33-2A3 (CF-Blue), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13723

Catalog No. 89-033-788


Add to cart

Description

Description

Mouse monoclonal antibody raised against human CD3E.

  • High specificity
  • High reactivity
Specifications

Specifications

CD3E
33-2A3
Mouse monoclonal antibody raised against human CD3E.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
CD3E
Human leukocytes.
Protein A/G purification
RUO
Primary
916
Flow Cytometry
Unconjugated
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
P07766
FLJ18683|T3E|TCRE
Mouse
IgG2a
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)