Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3E Mouse anti-Human, Clone: C3e/1308, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14702

Catalog No. 89-034-785


Add to cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human CD3E.

  • High specificity
  • High reactivity
Specifications

Specifications

CD3E
C3e/1308
Mouse monoclonal antibody raised against partial recombinant human CD3E.
In 10 mM PBS.
FLJ18683|T3E|TCRE
Mouse
IgG2b, kappa
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CD3E
Recombinant protein corresponding to amino acids 23-119 of human CD3E.
Protein A purification
RUO
Primary
916
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)