Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3e Rabbit anti-Human,Monkey, KHL, Clone: 1H4, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen CD3e
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB101125MI
View Documents
MilliporeSigma
ZRB101125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10114XMI
View Documents
MilliporeSigma
ZRB10114X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD3e Monoclonal antibody specifically detects CD3e in Human,Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

CD3e
KHL
Rabbit
T-cell surface glycoprotein CD3 epsilon chain;CD3-epsilon;CD3e;T-cell surface antigen T3/Leu-4 epsilon chain
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human T-cell surface glycoprotein CD3 epsilon chain (CD3e).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Monkey
CD3E, T3E
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit