Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, anti-Human, Clone: YNB46.1.8 (Campath-9, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00155

Catalog No. 89-032-498


Add to cart

Specifications

Specifications

CD4
YNB46.1.8 (Campath-9
Flow Cytometry (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
CD4 molecule
CD4mut
Original antibody is raised against rat T-cell line (NB2) transfected with the human CD4.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
920
Human
Flow Cytometry, Immunohistochemistry
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
CD4
IgG1, kappa
RUO
Antibody
Monoclonal
This antibody recognizes 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit