Learn More
CD4/CD28 Antibody Cocktail, FITC, PE, Exalpha

Mouse Cocktail Antibody
Supplier: Vector Laboratories 0428S


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD4/CD28 Cocktail antibody specifically detects CD4/CD28 in Human samples. It is validated for Flow CytometrySpecifications
CD4/CD28 | |
Cocktail | |
PBS with 0.08% sodium azide | |
P01730, P10747 | |
Mouse | |
Protein A/G | |
RUO | |
920, 940 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry | |
PE | |
CD28 | |
CD28; CD28 antigen; CD28 molecule; CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; MGC138290; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell-specific surface glycoprotein CD28; Tp44 | |
CD4 was derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human perherial blood T lymphocytes.CD28 was derived from the hybridization of mouse Sp2/O-Ag14 myeloma cells with spleen cells of BALB/c mice immunized with the HPB-ALL T-cell line. | |
50Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.