Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Human anti-Human, Primate, DyLight™ 680, Clone: CE9.1 (Clenoliximab), Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275910FR

Catalog No. NBP275910FR


Add to cart

Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human, Primate samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.
Specifications

Specifications

CD4
CE9.1 (Clenoliximab)
Flow Cytometry, ELISA, Immunohistochemistry
Conjugated, Purified
CD4
Domain 1 of human CD4.
Protein A purified
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers
Primary
920.0
Human, Primate
ELISA, Flow Cytometry, Immunohistochemistry
DyLight 680
PBS with 0.05% Sodium Azide
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Human
IgG4 Lambda
0.1 mL
Store at 4°C in the dark.
Monoclonal
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit