Learn More
CD4 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: L200, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566479
 View more versions of this product
Catalog No. BDB566479
Add to cart
Description
Description
CD4 Monoclonal antibody specifically detects CD4 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD4
L200
BB700
Mouse
0.1 mg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Baboon, Cynomolgus, Rhesus Macaque, Human