Learn More
CD4 Mouse anti-Human, Alexa Fluor 405, Clone: 11830, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems FAB3791V025
Catalog No. FAB3791V025
Add to cart
This item is not returnable. View return policy
Description
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD4
11830
Flow Cytometry 5 uL/10^6 cells
P01730
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CD4 in direct ELISAs.
Flow Cytometry
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CD4
Recombinant human CD4 extracellular domain Accession # P01730
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
920
Human