Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human , BUV615, Clone: SK3 (also known as Leu3a), BD Biosciences™

Mouse Monoclonal Antibody

$227.00 - $437.50

Specifications

Antigen CD4
Clone SK3 (also known as Leu3a)
Applications Flow Cytometry
Conjugate Brilliant Ultraviolet 615
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612988
View Documents
BD Biosciences
612988
25 Tests
Each for $227.00
Add to cart
 
BDB612987
View Documents
BD Biosciences
612987
100 Tests
Each for $437.50
Add to cart
 
Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD4
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Human Peripheral Blood T Cells
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
SK3 (also known as Leu3a)
Brilliant Ultraviolet 615
Mouse
Human
L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55)
Mouse BALB/c IgG1, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit