Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human , BV510, Clone: OKT4, BD Biosciences™

Mouse Monoclonal Antibody

$177.10 - $393.00

Specifications

Antigen CD4
Clone OKT4
Applications Flow Cytometry
Conjugate Brilliant Violet 510
Format Conjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566805
View Documents
BD Biosciences
566805
25 Tests
Each for $177.10
Add to cart
 
BDB566804
View Documents
BD Biosciences
566804
100 Tests
Each for $393.00
Add to cart
 
Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD4
Flow Cytometry
Conjugated
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Sheep Erythrocyte Rosette-purified Human T Cells
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed.
OKT4
Brilliant Violet 510
Mouse
Human
CD4 antigen (p55); T-cell surface antigen T4/Leu-3
Mouse BALB/c x A/J, also known as CAF1 IgG2b, κ
Primary
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit